Learn More
MSC Ironclad Cut A4 Gloves, Puncture 3, Large, 1 Pair
Catalog No. 5000688920
Encompass
Ironclad Cut, Puncture and Abrasion Resistant Gloves are designed for mechanic's style applications requiring high levels of protection. These gloves feature an HPPE steel blend knit and a foam nitrile-coated palm, providing excellent cut, puncture, and abrasion resistance.
  • Foam nitrile Command™ touchscreen coated palm
  • HPPE/Steel blended knit construction, 13 gauge
  • Color coded thumb saddle reinforcement
  • Cut resistance level A4
  • Puncture resistance level 3
  • Abrasion resistance level 3
  • Seamless lining construction
  • Foam grip surface
  • Knit cuff style
Catalog No. 50-006-88920 Supplier MSC Supplier No. 20598306
Ironclad Cut, Puncture and Abrasion Resistant Gloves are designed for mechanic's style applications requiring high levels of protection. These gloves feature an HPPE steel blend knit and a foam nitrile-coated palm, providing excellent cut, puncture, and abrasion resistance.
  • Foam nitrile Command™ touchscreen coated palm
  • HPPE/Steel blended knit construction, 13 gauge
  • Color coded thumb saddle reinforcement
  • Cut resistance level A4
  • Puncture resistance level 3
  • Abrasion resistance level 3
  • Seamless lining construction
  • Foam grip surface
  • Knit cuff style