Learn More
MSC Ironclad Cut Gloves, A6, Foam Nitrile, Touchscreen, 12 Pairs
Catalog No. 5000789714
Encompass
The COMMAND™ A6 Foam Nitrile gloves are engineered for robust hand protection in environments requiring high cut resistance and tactile dexterity. Featuring a 13 gauge HPPE/steel blended knit, these gloves offer superior performance against cuts and punctures, alongside touchscreen compatibility for seamless operation of devices.
  • Touchscreen conductive palm coating
  • Nitrile reinforced thumb saddle
  • Color-coded to indicate cut protection level
  • ANSI A6 cut resistance and ANSI 4 puncture resistance
  • Individually wrapped for hygiene and convenience
Catalog No. 50-007-89714 Supplier MSC Supplier No. 16666554
The COMMAND™ A6 Foam Nitrile gloves are engineered for robust hand protection in environments requiring high cut resistance and tactile dexterity. Featuring a 13 gauge HPPE/steel blended knit, these gloves offer superior performance against cuts and punctures, alongside touchscreen compatibility for seamless operation of devices.
  • Touchscreen conductive palm coating
  • Nitrile reinforced thumb saddle
  • Color-coded to indicate cut protection level
  • ANSI A6 cut resistance and ANSI 4 puncture resistance
  • Individually wrapped for hygiene and convenience