Learn More
MSC IronClad Cut-Resistant Gloves, A5 Cut, 13 Gauge, 12 Each
Catalog No. 5000676157
Encompass
These gloves are cut, puncture, and abrasion-resistant, designed for tasks requiring robust hand protection. They feature an HPPE steel blend knit with a nitrile-coated, 3/4 dip and palm coating for enhanced durability and grip, suitable for various demanding applications.
  • Cut, puncture, and abrasion-resistant design
  • Primary material: HPPE steel blend knit
  • Nitrile coating for enhanced grip and durability
  • ANSI/ISEA cut resistance level A5
  • ANSI/ISEA abrasion resistance level 4
  • ANSI/ISEA puncture resistance level 4
  • Flexible 13 gauge knit
  • Touchscreen compatible palm coating
  • Seamless HPPE blend lining
  • Meets ANSI 105 and EN388 2016 standards
  • Double coated style
  • Package quantity: 12
Catalog No. 50-006-76157 Supplier MSC Supplier No. 16685646
These gloves are cut, puncture, and abrasion-resistant, designed for tasks requiring robust hand protection. They feature an HPPE steel blend knit with a nitrile-coated, 3/4 dip and palm coating for enhanced durability and grip, suitable for various demanding applications.
  • Cut, puncture, and abrasion-resistant design
  • Primary material: HPPE steel blend knit
  • Nitrile coating for enhanced grip and durability
  • ANSI/ISEA cut resistance level A5
  • ANSI/ISEA abrasion resistance level 4
  • ANSI/ISEA puncture resistance level 4
  • Flexible 13 gauge knit
  • Touchscreen compatible palm coating
  • Seamless HPPE blend lining
  • Meets ANSI 105 and EN388 2016 standards
  • Double coated style
  • Package quantity: 12