Learn More
MSC IronClad General Purpose Gloves, WCG, Medium, Terry/Suede Grip, 1 Pair
Catalog No. 5000787004
Encompass
These ironClad general purpose gloves are designed for comfort and durability in demanding work environments. Featuring a terrycloth lining and a suede grip, they are suitable for various applications including power sports, professional mechanics, and hand & power tool use.
  • High performance synthetic suede palm for balanced comfort and durability
  • Breathable nylon back of hand offers durable comfort
  • Neoprene compression cuff
  • TPR cuff puller allows for easy on and off
  • Thumb saddle reinforcement increases durability
  • Terry cloth sweat wipe for managing face sweat
Catalog No. 50-007-87004 Supplier MSC Supplier No. 99493843
These ironClad general purpose gloves are designed for comfort and durability in demanding work environments. Featuring a terrycloth lining and a suede grip, they are suitable for various applications including power sports, professional mechanics, and hand & power tool use.
  • High performance synthetic suede palm for balanced comfort and durability
  • Breathable nylon back of hand offers durable comfort
  • Neoprene compression cuff
  • TPR cuff puller allows for easy on and off
  • Thumb saddle reinforcement increases durability
  • Terry cloth sweat wipe for managing face sweat