Learn More
MSC IronClad Gloves, A4 Cut, Puncture 5, Small, 12 Pair
Catalog No. 5000688526
Encompass
These mechanic's style gloves are engineered for robust protection, offering superior cut and puncture resistance for demanding industrial applications. They feature an HPPE steel blend knit with a smooth nitrile palm coating.
  • Cut and puncture-resistant design
  • HPPE steel blend knit construction for durability
  • Nitrile-coated palm for enhanced grip
  • Touchscreen compatibility for convenience
  • Seamless lining for comfort
  • Knit cuff for a secure fit
Catalog No. 50-006-88526 Supplier MSC Supplier No. 97152045
These mechanic's style gloves are engineered for robust protection, offering superior cut and puncture resistance for demanding industrial applications. They feature an HPPE steel blend knit with a smooth nitrile palm coating.
  • Cut and puncture-resistant design
  • HPPE steel blend knit construction for durability
  • Nitrile-coated palm for enhanced grip
  • Touchscreen compatibility for convenience
  • Seamless lining for comfort
  • Knit cuff for a secure fit