Learn More
MSC IronClad Gloves, ANSI Cut A2, Puncture 3, 1 Pair
Catalog No. 5000677708
Encompass
These gloves are designed for high resistance to cuts, punctures, and abrasions. Constructed from synthetic leather with a suede grip, they offer robust protection and comfort for demanding applications.
  • ANSI Cut A2
  • ANSI Puncture 3
  • ANSI Abrasion 3
  • Large size
  • Synthetic leather material
  • Black color
  • Suede grip surface
  • Water-resistant Command touchscreen synthetic suede palm
  • Neoprene back of hand
  • TPR knuckle impact protection
  • Thumb saddle reinforcement
Catalog No. 50-006-77708 Supplier MSC Supplier No. 10433555
These gloves are designed for high resistance to cuts, punctures, and abrasions. Constructed from synthetic leather with a suede grip, they offer robust protection and comfort for demanding applications.
  • ANSI Cut A2
  • ANSI Puncture 3
  • ANSI Abrasion 3
  • Large size
  • Synthetic leather material
  • Black color
  • Suede grip surface
  • Water-resistant Command touchscreen synthetic suede palm
  • Neoprene back of hand
  • TPR knuckle impact protection
  • Thumb saddle reinforcement