Learn More
MSC Ironclad Gloves, ANSI Cut A6, Impact, X-Large, 1 Pair
Catalog No. 5000802430
Encompass
Ironclad Command Impact Cut A6 Gloves provide advanced protection against cuts and impacts, designed for demanding work environments. These gloves feature a durable construction with enhanced dexterity, making them suitable for various industrial applications.
  • TPR impact protection on fingers, thumb, and knuckle
  • Cut-resistant 360° A6 HPPE blend liner
  • Reinforced touchscreen synthetic suede padded palm
  • Breathable polyester knit back of hand
  • Thumb saddle reinforcement for extended durability
  • Padded palm for hard use and comfort
  • Touchscreen compatible for modern applications
  • Velcro closure for easy entry and secure fit
  • Terry cloth sweat wipe thumb
Catalog No. 50-008-02430 Supplier MSC Supplier No. 13859376
Ironclad Command Impact Cut A6 Gloves provide advanced protection against cuts and impacts, designed for demanding work environments. These gloves feature a durable construction with enhanced dexterity, making them suitable for various industrial applications.
  • TPR impact protection on fingers, thumb, and knuckle
  • Cut-resistant 360° A6 HPPE blend liner
  • Reinforced touchscreen synthetic suede padded palm
  • Breathable polyester knit back of hand
  • Thumb saddle reinforcement for extended durability
  • Padded palm for hard use and comfort
  • Touchscreen compatible for modern applications
  • Velcro closure for easy entry and secure fit
  • Terry cloth sweat wipe thumb