Learn More
MSC IronClad Gloves, COMMAND IMPACT CUT A6, Small, 1 Pair
Catalog No. 5000685199
Encompass
These versatile work gloves are engineered for superior protection in demanding job site conditions, offering advanced resistance to cuts, punctures, and abrasions, alongside comprehensive impact protection.
  • Full 360° cut A6 HPPE blend liner
  • TPR impact protection for fingers, thumb, and knuckle
  • Reinforced and padded synthetic suede palm with touchscreen compatibility
  • Breathable polyester knit back of hand
  • Thumb saddle reinforcement
  • Velcro closure for easy entry and secure fit
  • Terry cloth sweat wipe thumb
Catalog No. 50-006-85199 Supplier MSC Supplier No. 13858931
These versatile work gloves are engineered for superior protection in demanding job site conditions, offering advanced resistance to cuts, punctures, and abrasions, alongside comprehensive impact protection.
  • Full 360° cut A6 HPPE blend liner
  • TPR impact protection for fingers, thumb, and knuckle
  • Reinforced and padded synthetic suede palm with touchscreen compatibility
  • Breathable polyester knit back of hand
  • Thumb saddle reinforcement
  • Velcro closure for easy entry and secure fit
  • Terry cloth sweat wipe thumb