Learn More
MSC IronClad Gloves, Cut A1, Puncture 2, Medium, 1 Pair
Catalog No. 5000796297
Encompass
The Command Series Touchscreen gloves from ironClad use nanoparticle technology with highly conductive carbon ions, offering electrical conductivity superior to a human finger. Their specialized ion-infused microfiber matrix ensures long-lasting performance, ideal for modern workplaces requiring quality, durability, and a flawless fit for use with tablets, data entry terminals, and smartphones.
  • Touchscreen-conductive palm and fingers for seamless device interaction
  • Adjustable hook and loop closure ensures secure, custom fit
  • Silicone-fused palm provides maximum grip, resists peeling
  • Neoprene knuckle feature for enhanced impact and abrasion protection
  • Machine washable, resists shrinking or drying out
  • Thumb saddle reinforcement for increased durability
  • Terry cloth sweat wipe for managing sweat
Catalog No. 50-007-96297 Supplier MSC Supplier No. 10433084
The Command Series Touchscreen gloves from ironClad use nanoparticle technology with highly conductive carbon ions, offering electrical conductivity superior to a human finger. Their specialized ion-infused microfiber matrix ensures long-lasting performance, ideal for modern workplaces requiring quality, durability, and a flawless fit for use with tablets, data entry terminals, and smartphones.
  • Touchscreen-conductive palm and fingers for seamless device interaction
  • Adjustable hook and loop closure ensures secure, custom fit
  • Silicone-fused palm provides maximum grip, resists peeling
  • Neoprene knuckle feature for enhanced impact and abrasion protection
  • Machine washable, resists shrinking or drying out
  • Thumb saddle reinforcement for increased durability
  • Terry cloth sweat wipe for managing sweat