Learn More
MSC IronClad Gloves, Cut A4, 2X-Large, Foam Nitrile Palm, 1 ea
Catalog No. 5000688564
Encompass
These mechanic's style gloves are engineered for high-performance hand protection, offering superior cut, puncture, and abrasion resistance. Constructed with an HPPE Steel Blend Knit and featuring a foam nitrile-coated palm, they provide both durability and enhanced grip for demanding tasks.
  • Cut resistance level A4 (ANSI/ISEA)
  • Puncture resistance level 3 (ANSI/ISEA)
  • Abrasion resistance level 3 (ANSI/ISEA)
  • Made with HPPE steel blend knit
  • Foam nitrile-coated palm for enhanced grip
  • Touchscreen compatible palm coating
  • Features 13 gauge construction
  • Provided in 2X-Large size (numeric size 11)
  • Knit cuff style
  • Complies with ANSI 105 and EN388 2016 standards
Catalog No. 50-006-88564 Supplier MSC Supplier No. 97166672
These mechanic's style gloves are engineered for high-performance hand protection, offering superior cut, puncture, and abrasion resistance. Constructed with an HPPE Steel Blend Knit and featuring a foam nitrile-coated palm, they provide both durability and enhanced grip for demanding tasks.
  • Cut resistance level A4 (ANSI/ISEA)
  • Puncture resistance level 3 (ANSI/ISEA)
  • Abrasion resistance level 3 (ANSI/ISEA)
  • Made with HPPE steel blend knit
  • Foam nitrile-coated palm for enhanced grip
  • Touchscreen compatible palm coating
  • Features 13 gauge construction
  • Provided in 2X-Large size (numeric size 11)
  • Knit cuff style
  • Complies with ANSI 105 and EN388 2016 standards