Learn More
MSC IronClad Gloves, Cut A4, Puncture 3, Abrasion 3, X-Large, 12/Pk.
Catalog No. 5000676712
Encompass
These cut, puncture, and abrasion-resistant gloves feature an HPPE steel blend knit with a foam nitrile-coated palm, providing enhanced grip and protection for various industrial applications.
  • Mechanic's style
  • Foam nitrile coating material with palm coverage
  • ANSI/ISEA A4 cut resistance, level 3 puncture resistance, and level 3 abrasion resistance
  • Primary material is HPPE steel blend knit, with HPPE blend back and seamless lining
  • X-Large size with a 13 gauge knit
  • Touchscreen compatible palm
  • EN 388 rating of 4X42D
  • Standards include ANSI 105 and EN388 2016
  • Foam grip surface
  • Knit cuff style
  • Sold in a package quantity of 12
Catalog No. 50-006-76712 Supplier MSC Supplier No. 97167308
These cut, puncture, and abrasion-resistant gloves feature an HPPE steel blend knit with a foam nitrile-coated palm, providing enhanced grip and protection for various industrial applications.
  • Mechanic's style
  • Foam nitrile coating material with palm coverage
  • ANSI/ISEA A4 cut resistance, level 3 puncture resistance, and level 3 abrasion resistance
  • Primary material is HPPE steel blend knit, with HPPE blend back and seamless lining
  • X-Large size with a 13 gauge knit
  • Touchscreen compatible palm
  • EN 388 rating of 4X42D
  • Standards include ANSI 105 and EN388 2016
  • Foam grip surface
  • Knit cuff style
  • Sold in a package quantity of 12