Learn More
MSC IronClad Gloves, Cut A6, Puncture 4, Abrasion 4, 1 pair.
Catalog No. 5000686745
Encompass
These gloves are engineered with an HPPE steel blend knit and a 3/4 dip and palm nitrile coating for superior cut, puncture, and abrasion resistance. Featuring a sandy grip surface and touchscreen palm, they are designed for demanding applications.
  • Cut, puncture, and abrasion-resistant design
  • HPPE steel blend knit material for durability
  • Nitrile-coated with 3/4 dip and palm coverage
  • Sandy grip surface enhances handling
  • Touchscreen compatible palm for convenience
  • ANSI/ISEA cut resistance level A6
  • ANSI/ISEA puncture resistance level 4
  • ANSI/ISEA abrasion resistance level 4
  • Seamless knit construction, 13-gauge
  • Double coated for enhanced durability
  • Complies with ANSI 105 and EN388 2016 standards
Catalog No. 50-006-86745 Supplier MSC Supplier No. 16685653
These gloves are engineered with an HPPE steel blend knit and a 3/4 dip and palm nitrile coating for superior cut, puncture, and abrasion resistance. Featuring a sandy grip surface and touchscreen palm, they are designed for demanding applications.
  • Cut, puncture, and abrasion-resistant design
  • HPPE steel blend knit material for durability
  • Nitrile-coated with 3/4 dip and palm coverage
  • Sandy grip surface enhances handling
  • Touchscreen compatible palm for convenience
  • ANSI/ISEA cut resistance level A6
  • ANSI/ISEA puncture resistance level 4
  • ANSI/ISEA abrasion resistance level 4
  • Seamless knit construction, 13-gauge
  • Double coated for enhanced durability
  • Complies with ANSI 105 and EN388 2016 standards