Learn More
MSC IronClad Gloves, High-Visibility, ANSI Cut A6, Large, 1 Pair
Catalog No. 5000766379
Encompass
These gloves provide high-visibility impact protection and offer excellent resistance against cuts, punctures, and abrasions. They are designed for demanding work environments, featuring a padded palm and robust construction for enhanced hand safety.
  • High-visibility orange and yellow design
  • Padded synthetic suede palm for grip and comfort
  • Breathable polyester knit back of hand
  • Liner is 360° cut A5 HPPE blend
  • TPR back of hand and thumb impact protection
  • Thumb saddle reinforcement for durability
  • Hook & loop cuff closure
  • ANSI Cut A6, ANSI Puncture 4, ANSI Abrasion 4, ANSI Impact 1 rated
Catalog No. 50-007-66379 Supplier MSC Supplier No. 97168488
These gloves provide high-visibility impact protection and offer excellent resistance against cuts, punctures, and abrasions. They are designed for demanding work environments, featuring a padded palm and robust construction for enhanced hand safety.
  • High-visibility orange and yellow design
  • Padded synthetic suede palm for grip and comfort
  • Breathable polyester knit back of hand
  • Liner is 360° cut A5 HPPE blend
  • TPR back of hand and thumb impact protection
  • Thumb saddle reinforcement for durability
  • Hook & loop cuff closure
  • ANSI Cut A6, ANSI Puncture 4, ANSI Abrasion 4, ANSI Impact 1 rated