Learn More
Ironclad Performance Wear Ironclad Heavy Utility Glove, 12 pr per box, size Mediuml, Black/grey, EN388
Catalog No. 501610041
Encompass_Preferred
Gloves, Ironclad, Heavy Utility Glove, High abrasion resistance, Breathable nylon back of hand, Duraclad reinforced palm, Knuckle impact TPR, Hybrid safety cuff. Ideal for construction, material handling, demolition, search and rescue, landscaping, forestry, lumberyards, cable and wire work.12 pair per box
This item is not returnable. View return policy
Catalog No. 50-161-0041 Supplier Ironclad Performance Wear Supplier No. HUG03M
Gloves, Ironclad, Heavy Utility Glove, High abrasion resistance, Breathable nylon back of hand, Duraclad reinforced palm, Knuckle impact TPR, Hybrid safety cuff. Ideal for construction, material handling, demolition, search and rescue, landscaping, forestry, lumberyards, cable and wire work.12 pair per box