Learn More
MSC IronClad IEX-HVP Gloves, ANSI Cut A2/Puncture 5, X-Large, 1 Pair
Catalog No. 5000796470
Encompass
These ironClad gloves, model IEX-HVP, are designed for protection against cuts, punctures, and abrasions. They are suitable for various industrial applications requiring high visibility and hand safety.
  • Command™ touchscreen synthetic suede palm
  • Breathable polyester knit back of hand
  • Neoprene knuckle impact protection
  • Thumb saddle reinforcement
  • Terry cloth sweat wipe
  • Hook & loop cuff style
  • High visibility design
  • Full fingered
  • Uncoated coating material and coverage
Catalog No. 50-007-96470 Supplier MSC Supplier No. 10432938
These ironClad gloves, model IEX-HVP, are designed for protection against cuts, punctures, and abrasions. They are suitable for various industrial applications requiring high visibility and hand safety.
  • Command™ touchscreen synthetic suede palm
  • Breathable polyester knit back of hand
  • Neoprene knuckle impact protection
  • Thumb saddle reinforcement
  • Terry cloth sweat wipe
  • Hook & loop cuff style
  • High visibility design
  • Full fingered
  • Uncoated coating material and coverage