Learn More
Ironclad Performance Wear Ironclad Workforce Glove, 12 pr per box, size Small, Black
Catalog No. 501610064
Encompass_Preferred
Gloves, Ironclad, Workforce, Slip-on all purpose glove, Breathable nylon back of hand, Microsuede palm, Knuckle TPR impact protection, Fingernail guards. Ideal for Home, shop, light construction, Maintenance, Landscaping, Yard work. 12 pr per box.
This item is not returnable. View return policy
Catalog No. 50-161-0064 Supplier Ironclad Performance Wear Supplier No. WFG02S
Gloves, Ironclad, Workforce, Slip-on all purpose glove, Breathable nylon back of hand, Microsuede palm, Knuckle TPR impact protection, Fingernail guards. Ideal for Home, shop, light construction, Maintenance, Landscaping, Yard work. 12 pr per box.