Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI Iron(II) Acetate, Reagent, 90.0% Purity, 25 g

Catalog No. 502781476
Encompass_Preferred

Iron(II) Acetate is a reagent-grade inorganic chemical compound frequently utilized in industrial applications, such as acting as a mordant within the dye industry. This product is formulated to support research and industrial chemical processes.

  • Formula: C4H6O4Fe
  • Molecular weight: 173.93
  • Purity: ≥90.0% (potassium permanganate method)
  • Physical state: Solid (white to gray to brown powder or crystal)
  • Storage: Room temperature, preferably in a cool, dark place (<15°C) under inert gas
  • Sensitivity: Air sensitive

Catalog No. 50-278-1476 Supplier Grainger Supplier No. 19KK12
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iron(II) Acetate is a reagent-grade inorganic chemical compound frequently utilized in industrial applications, such as acting as a mordant within the dye industry. This product is formulated to support research and industrial chemical processes.

  • Formula: C4H6O4Fe
  • Molecular weight: 173.93
  • Purity: ≥90.0% (potassium permanganate method)
  • Physical state: Solid (white to gray to brown powder or crystal)
  • Storage: Room temperature, preferably in a cool, dark place (<15°C) under inert gas
  • Sensitivity: Air sensitive

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.