Learn More
Sigma Aldrich Fine Chemicals Biosciences IronII sulfate heptahydrat1KG
Catalog No. 502898825
Encompass
Iron(II) sulfate heptahydrate is a heptahydrate of iron(II) sulfate. It undergoes thermal degradation to afford hydrosulfate and oxysulfate of iron(III). Isothermal and dynamic thermal analysis methods have been proposed to study its thermal decomposition in air.
Catalog No. 50-289-8825 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 123541KG
Iron(II) sulfate heptahydrate is a heptahydrate of iron(II) sulfate. It undergoes thermal degradation to afford hydrosulfate and oxysulfate of iron(III). Isothermal and dynamic thermal analysis methods have been proposed to study its thermal decomposition in air.