Learn More
Sigma Aldrich Fine Chemicals Biosciences IronII sulfate heptahydrat5KG
Catalog No. 502902976
Encompass
Iron(II) sulfate heptahydrate is a heptahydrate of iron(II) sulfate. It undergoes thermal degradation to afford hydrosulfate and oxysulfate of iron(III). Isothermal and dynamic thermal analysis methods have been proposed to study its thermal decomposition in air.
Catalog No. 50-290-2976 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 123545KG
Iron(II) sulfate heptahydrate is a heptahydrate of iron(II) sulfate. It undergoes thermal degradation to afford hydrosulfate and oxysulfate of iron(III). Isothermal and dynamic thermal analysis methods have been proposed to study its thermal decomposition in air.