Learn More
Sigma Aldrich Fine Chemicals Biosciences IronIII chloride hexahydra1KG
Catalog No. 502898251
Encompass
Iron(III) chloride hexahydrate can be obtained from crystallization of iron(III) chloride from water. Its crystal structure has been studied using three-dimensional single-crystal X-ray-diffraction data. Its crystals have two chloride ions and four water molecules arranged around each ferric ion to form octahedral trans-[feCl2(OH2)4] ions having C2h-2/m symmetry.
Catalog No. 50-289-8251 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 312321KG
Iron(III) chloride hexahydrate can be obtained from crystallization of iron(III) chloride from water. Its crystal structure has been studied using three-dimensional single-crystal X-ray-diffraction data. Its crystals have two chloride ions and four water molecules arranged around each ferric ion to form octahedral trans-[feCl2(OH2)4] ions having C2h-2/m symmetry.