Learn More
Selleck Chemical LLC Irosustat
SDP
Catalog No. 5000000584
Encompass_Preferred
Irosustat (STX64 BN83495) a tricyclic coumarin sulfamate is an orally active nonsteroid-based irreversible steroid sulfatase inhibitor with an IC50 of 8 nM
Catalog No. 50-000-00584 Supplier Selleck Chemical LLC Supplier No. S65585MG
Irosustat (STX64 BN83495) a tricyclic coumarin sulfamate is an orally active nonsteroid-based irreversible steroid sulfatase inhibitor with an IC50 of 8 nM