Learn More
Medchemexpress LLC HY-14586 5mg Medchemexpress, Irosustat CAS:288628-05-7 Purity:>98%
SDP
Catalog No. 502081617
Encompass_Preferred
Medchemexpress, HY-14586 5mg Irosustat CAS:288628-05-7 Irosustat is a potent steroid sulfatase inhibitor, with an IC50 of 8 nM, and exhibits anti-breast cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1617 Supplier Medchemexpress LLC Supplier No. HY145865MG
Medchemexpress, HY-14586 5mg Irosustat CAS:288628-05-7 Irosustat is a potent steroid sulfatase inhibitor, with an IC50 of 8 nM, and exhibits anti-breast cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.