Learn More
Benchmark Products Inc Benchmark Products Inc. IRR Clean Polybags, Gamma-Irradiated, 10⁻⁶ SAL, 33 x 40 in
SDP
Catalog No. NC9908886
Encompass_Preferred
The Benchmark Products Inc. PB3340-4-IR are gamma-irradiated, sterile cleanroom bags designed for critical environments. Measuring 33x40 in and featuring a 4 mil thickness, these high-clarity polyethylene bags provide a 10⁻⁶ Sterility Assurance Level for exceptional contamination prevention. Their double-bagged design and lot number traceability ensure the highest product integrity during aseptic transfers, making them a superior choice for rigorous cleanroom applications.
Catalog No. NC9908886 Supplier Benchmark Products Inc Supplier No. PB33404IR
The Benchmark Products Inc. PB3340-4-IR are gamma-irradiated, sterile cleanroom bags designed for critical environments. Measuring 33x40 in and featuring a 4 mil thickness, these high-clarity polyethylene bags provide a 10⁻⁶ Sterility Assurance Level for exceptional contamination prevention. Their double-bagged design and lot number traceability ensure the highest product integrity during aseptic transfers, making them a superior choice for rigorous cleanroom applications.