Learn More
Contec™ Irradiated Bucket Bag Liners
BIOPHARMAGREENER_CHOICE
Catalog No. 19169125 Shop All Contec Inc. Products
Change view
Click to view available options
Capacity (English):
3.9 gal
Catalog No. Capacity (English)
19-169-125 3.9 gal
1 options
Catalog No. 19-169-125 Supplier Contec™ Supplier No. BKTBAG2
Add to Cart
Edge
Add to Cart
Contec™ Irradiated Bucket Bag Liners are for use with Contec™ 15 L polypropylene buckets.
  • Protect buckets and disinfectant solutions by reducing the opportunity for cross contamination
  • Intended for single use
  • Provide users with no access to autoclaves the ability to keep solutions sterile
Specifications
Product Type Bucket Liner
Material Polyethylene
Autoclavable Not Autoclavable
Capacity (English) 3.9 gal
Disposable Single-use
For Use With (Application) Cleaning and disinfecting
For Use With (Equipment) Contec 15 L Polypropylene Bucket
Quantity 3/Bag, 20 Bags/Cs.
Sterility Sterile