Learn More
Thermo Scientific™ Irradiated Isolator Wrap™ Sterile D/E (Dey-Engley) Neutralizing Agar Contact Plate
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Remel Irradiated Isolator Wrap Sterile D/E Neutralizing Agar Contact Plate, Irradiated is used for cultivation of microorganisms with neutralization of specific chemical disinfectants.
- Sterile Contact Plates are for surface sampling in isolator environments
- These products are manufactured in a clean room environment and validated sterile by gamma radiation
- The contact plates listed are made using GripFit™ technology, which ensures the lid remains in place during handling
- The plates are shrink-wrapped and contained in a protective outer barrier wrap, which is impermeable to isolator decontamination procedures but maintains the integrity of the medium inside
- An extra sterile bag is provided for transporting the plates to the laboratory for testing
Specifications
Specifications
| Format | Foil Barrier Wrap + Bag |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
Not intended for IVD use.