Description
- Blue twisted loop microdenier mop cover
- Microfiber string is made of polyester and nylon fibers for exceptional surface cleaning
- Three layer absorbency
- Individually wrapped
- Autoclavable
- Gamma irradiated
Specifications
Specifications
| Product Type | Mop Cover |
| Color | Blue |
| Sterility | Sterile |
| Autoclavable | Autoclavable |
| Color-coded | No |
| Material | Polyester, Nylon |