Learn More
Berkshire™ Irradiated V Foam Mop Cover
Catalog No. 19266706 Shop All Berkshire Corporation Products
Change view
Click to view available options
Color:
White, Blue
Catalog No. Color
19-266-706 White, Blue
1 options
This item is not returnable. View return policy
Catalog No. 19-266-706 Supplier Berkshire™ Supplier No. FMC6LF24IR
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mop cover constructed of polyester knit clean room fabric laminated to absorbent polyurethane foam.
  • Abrasion resistant polyester knit clean room fabric laminated to absorbent polyurethane foam
  • Laser cut to minimize fiber and particle release
  • 10 mm thickness provides cushioned surface for optimum conformance to uneven floors, walls and ceilings
  • High absorbency capacity
  • Compatible with a wide range of chemicals, disinfectants. and cleaners
  • Autoclavable and compatible with gamma and ETO sterilization
Specifications
Product Type Mop Cover
Color White, Blue
Sterility Sterile
Color-coded No
Material Polyurethane Foam