Learn More
ATCC IRR-MRC-5; Irradiated Fibroblast; Human (Homo sapiens)
Supplier:  ATCC 55X
Encompass
IRR-MRC-5 irradiated cells are a diploid cell culture line composed of fetal, lung fibroblasts. These cells are to be used as feeder cells to support the growth of other cells and as feeder cells to support the growth of stem cells in the undifferentiated state.
Catalog No. 50-238-1762