Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IRS1 (Phospho) [pS312] ELISA Kit

Catalog No. KHO0521
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KHO0521 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KHO0521 Supplier Invitrogen™ Supplier No. KHO0521
Add to Cart
Add to Cart

ELISA

The Human IRS1 (Phospho) [pS312] ELISA Kit is a solid-phase sandwich Enzyme-Linked Immunosorbent Assay (ELISA) designed to detect and quantify the level of IRS1 (Phospho) [pS312] in fresh or frozen human cell lysates. Cross-reactivity has been observed in mouse and rat cells. The assay recognizes both natural and recombinant IRS1 (Phospho) [pS312]. Principle of the method A monoclonal capture antibody specific for IRS1 (Phospho) [pS312] has been coated onto the wells of the 96-well plate. During the first incubation, standards of known content and unknown samples are pipetted into the wells and the antigen binds to the immobilized (capture) antibody. After washing, a rabbit antibody specific for the target protein is added to the wells and serves as a detection antibody by binding to the immobilized protein captured during the first incubation. After washing, a horseradish peroxidase labeled anti-rabbit IgG is added. This binds to the detection antibody to complete the four member sandwich. After a third incubation and washing to remove all the unbound enzyme, a substrate solution (TMB) is added, which is acted upon by the bound enzyme to produce color. The intensity of this colored product is directly proportional to the concentration of target protein present in the original specimen and the optical density can be read on a standard microplate reader. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for c...

IRS-1, a major substrate of the insulin receptor, is phosphorylated in response to stimulation of cells by insulin, insulin-like growth factor 1 (IGF-1) and interleukin 4 (IL-4). IRS-1 is phosphorylated on serine, threonine and tyrosine residues in a variety of tissues. An insulin-sensitive serine/threonine kinase casein kinase II mediates a portion of the insulin-stimulated serine/threonine phosphorylation of overexpressed IRS-1 in vivo. Thr 502 is identified as the major casein kinase II-catalyzed phosphorylation site in rat IRS-1, and Ser 99 is an additional phosphorylation site catalyzed by casein kinase II. Thus, casein kinase II-catalyzed phosphorylation of IRS-1 may be a component of the intracellular insulin signaling cascade. IRS-1 contains three putative binding sites for 14-3-3 (Ser 270, Ser 374 and Ser 641) and the motif around Ser 270 is located in the phosphortyrosine binding domain of IRS-1, which is responsible for the interaction with the insulin receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35568, P35569, P35570
Assay Range 1.6-100 U/mL
Assay Sensitivity <1 U/mL
Concentration 1.6 U/mL
Content And Storage Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Cell Lysates
Substrate Type Adaptor Proteins
Target Species Human, Mouse, Rat
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185; HIRS-1
Gene ID (Entrez) 16367, 25467, 3667
Gene Symbol IRS1
Incubation Time 4 hr.
Interassay CV 7.4%
Intraassay CV 5.8%
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Detection Antibody, anti-rabbit-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Standard Dilution Buffer, Detection Antibody, anti-rabbit-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene IRS-1
Protein Aliases IRS-1
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 10 μL
Shipping Condition Wet Ice
Storage Requirements 2-8°C
Target IRS1
Test Time 1 hr 20 min
Total Assay Time 4 hr
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.