Learn More
Apexbio Technology LLC Irsogladine 57381-26-7 50mg
SDP
Catalog No. 502240795
Encompass_Preferred
Irsogladine (CAS 57381-26-7) is a small molecule inhibitor of phosphodiesterase 4 (PDE4) and exhibits binding affinity for muscarinic acetylcholine receptors In preclinical studies irsogladine administration at 300 and 500 mg/kg/day produced a dose-dependent suppression of angiogenesis in wild-type mice reducing neovascularization by 21% and 45 3% respectively with statistical significance (P 0 02 P 0 001) These properties support its utility in investigating the modulation of cyclic AMP signaling cholinergic pathways and angiogenesis in biomedical research contexts
This item is not returnable. View return policy
Catalog No. 50-224-0795 Supplier Apexbio Technology LLC Supplier No. B159350
Irsogladine (CAS 57381-26-7) is a small molecule inhibitor of phosphodiesterase 4 (PDE4) and exhibits binding affinity for muscarinic acetylcholine receptors In preclinical studies irsogladine administration at 300 and 500 mg/kg/day produced a dose-dependent suppression of angiogenesis in wild-type mice reducing neovascularization by 21% and 45 3% respectively with statistical significance (P 0 02 P 0 001) These properties support its utility in investigating the modulation of cyclic AMP signaling cholinergic pathways and angiogenesis in biomedical research contexts