Learn More
Enzo Life Sciences Irsogladine. maleate (50mg). CAS: 84504-69-8
SDP
Catalog No. 502011380
Encompass_Preferred
Irsogladine is a selective PDE4 inhibitor which also enhances gap junction intercellular communication. It inhibits angiogenesis in wild-type and plasminogen activator-deficient mice. Purity: ≥98% (UHPLC). Solubility: Soluble in DMSO (25 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1380 Supplier Enzo Life Sciences Supplier No. BMLPD1410050
Irsogladine is a selective PDE4 inhibitor which also enhances gap junction intercellular communication. It inhibits angiogenesis in wild-type and plasminogen activator-deficient mice. Purity: ≥98% (UHPLC). Solubility: Soluble in DMSO (25 mg/ml). Long Term Storage: Ambient.