Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Irsogladine Maleate 98.0+%, TCI America™
SDP

Catalog No. I07191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07191G 1 g
I07195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07191G Supplier TCI America Supplier No. I07191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84504-69-8
Molecular Formula C13H11Cl2N5O4
Molecular Weight (g/mol) 372.16
MDL Number MFCD00873566
InChI Key PJLVTVAIERNDEQ-BTJKTKAUSA-N
Synonym irsogladine maleate, gaslon, gaslon n, unii-66gvu60epq, 66gvu60epq, 2,4-diamino-6-2,5-dichlorophenyl-s-triazine maleate, 6-2,5-dichlorophenyl-1,3,5-triazine-2,4-diamine maleate, 1,3,5-triazine-2,4-diamine, 6-2,5-dichlorophenyl-, z-2-butenedioate 1:1, dsstox_cid_20234
PubChem CID 5282435
IUPAC Name (2Z)-but-2-enedioic acid; 6-(2,5-dichlorophenyl)-1,3,5-triazine-2,4-diamine
SMILES OC(=O)\C=C/C(O)=O.NC1=NC(=NC(N)=N1)C1=CC(Cl)=CC=C1Cl

Specifications

Color White
Quantity 1 g
Formula Weight 372.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Irsogladine Maleate
RTECSNumber XY5930000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.