Learn More
Cayman Chemical Irsogladine (maleate), Purity: ≥98%, CAS: 84504-69-8, 50 mg
SDP
Catalog No. 502988329
Encompass_Preferred
A gastroprotective agent; increases transfer of Lucifer yellow CH between isolated rabbit gastric epithelial cells, indicating enhanced GJIC; inhibits gastric mucosal lesion formation and decreases in gastric mucosal blood flow induced by monochloramine in rats at 3 mg/kg, effects that can be prevented by L-NAME; inhibits fMLP-induced superoxide anion production and increases cAMP levels in isolated human neutrophils in a concentration-dependent manner, similar to rolipram
Catalog No. 50-298-8329 Supplier Cayman Chemical Supplier No. 3022350 MG
A gastroprotective agent; increases transfer of Lucifer yellow CH between isolated rabbit gastric epithelial cells, indicating enhanced GJIC; inhibits gastric mucosal lesion formation and decreases in gastric mucosal blood flow induced by monochloramine in rats at 3 mg/kg, effects that can be prevented by L-NAME; inhibits fMLP-induced superoxide anion production and increases cAMP levels in isolated human neutrophils in a concentration-dependent manner, similar to rolipram