Learn More
Air Boss™ Defense IRT Water Storage Bladders
Catalog No. 191300638 Shop All TVI Corporation Products
Click to view available options
Capacity (Metric):
346.3 L
5678 L
Capacity (English):
250 gal.
1500 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Capacity (English)
19-130-0638 5678 L 1500 gal.
19-130-0635 346.3 L 250 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-130-0638 Supplier Air Boss™ Defense Supplier No. WSB1500G
Includes
Protective ground cloth and transport bag
Effectively collect wastewater runoff from IRT decontamination shelters
  • Co-polymer material is highly resistant to chemicals, mold and mildew
  • Integral pressure relief valve, ball valve, back flow prevention valve, and cam-lok
  • Ball valve on either side of cam-lok for shut off, backflow prevention for one way fluid exchange, cam-lok for quick disconnect
  • Rugged design allows for use in the toughest environments
  • Quickly and easily deploys, no tools required, reusable and repairable
  • Compact and lightweight for portability, fully collapsible and foldable
Specifications
Product Type Waste Storage Bladder
Material Polyester
Capacity (English) 1500 gal.
Capacity (Metric) 5678 L
For Use With (Application) Collects waste water run-off from IRT decontamination shelters
Includes Protective ground Cloth and transport bag, integral pressure relief valve, ball valve, back flow prevention valve and cam-lok