Learn More
MSC IRWIN Aviation Snip, Straight/Wide Curve, 10 in, 1-5/16 in Cut
Catalog No. 5000509510
Encompass
This aviation snip features hot-drop forged steel blades for a sharper cutting edge and greater strength. Designed with ergonomically shaped ProTouch™ grips and a long-life spring, it enhances leverage and optimizes cuts. This 10 in snip has a 1-5/16 in length of cut, capable of straight and wide curve cuts, and can cut up to 18 gauge cold rolled steel and 23 gauge stainless steel.
  • Compound cutting action with textured grips and E-Z close latch
  • Great for cutting sheet metal, vinyl, plastic, rubber and many other applications
  • Multi-purpose snips with compound cutting leverage
  • Pro-Touch™ grips provide superior comfort, resist twisting
  • Hot-drop forged steel blades for sharper cutting edge and greater strength
  • Long-life spring enhances leverage and optimizes cuts
Catalog No. 50-005-09510 Supplier MSC Supplier No. 88531611
This aviation snip features hot-drop forged steel blades for a sharper cutting edge and greater strength. Designed with ergonomically shaped ProTouch™ grips and a long-life spring, it enhances leverage and optimizes cuts. This 10 in snip has a 1-5/16 in length of cut, capable of straight and wide curve cuts, and can cut up to 18 gauge cold rolled steel and 23 gauge stainless steel.
  • Compound cutting action with textured grips and E-Z close latch
  • Great for cutting sheet metal, vinyl, plastic, rubber and many other applications
  • Multi-purpose snips with compound cutting leverage
  • Pro-Touch™ grips provide superior comfort, resist twisting
  • Hot-drop forged steel blades for sharper cutting edge and greater strength
  • Long-life spring enhances leverage and optimizes cuts