Learn More
Aviva Systems Biology Corp Irx2 Antibody - C-terminal region (ARP37072_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37072_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Irx2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the c terminal region of mouse Irx2. Alternate names: IRX6, MGC103290ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-284-579