Learn More
Aviva Systems Biology Corp IRX3 Antibody - N-terminal region (ARP36905_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36905_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against IRX3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse IRX3. Alternate names: AI894186ApplicationWB. Species reactivity: Cow, Dog, Mouse, Pig, Rat
Catalog No. 50-014-56293