Learn More
Aviva Systems Biology Corp IRX3 Antibody - N-terminal region (ARP42800_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42800_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IRX3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IRX3. Alternate names: IRX-1, IRXB1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58338