Learn More
Aviva Systems Biology Corp IRX4 Antibody - N-terminal region (ARP32066_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32066_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against IRX4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IRX4. Alternate names: MGC131996, IRXA3ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rat
Catalog No. 50-014-54441