Learn More
Biomatik Corporation Irx4 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11175
Encompass_Preferred
Irx4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IRX4. The Irx4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Iroquois-Class Homeodomain Protein Irx-4 Protein (287-363Aa). Applications: ELISA, WB, IF. Target Synonyms: IRX4. Accession Number: P78413. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9250