Learn More
Aviva Systems Biology Corp Irx5 Antibody - middle region (ARP37245_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37245_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Irx5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of mouse Irx5. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-605