Learn More
Aviva Systems Biology Corp IRX6 Antibody - N-terminal region (ARP47679_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47679_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IRX6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IRX6. Alternate names: IRX-3, IRX7, IRXB3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59634