Learn More
Waters Corp Waters Quality Parts Solution, 10 mL, ISAAC Reference Electrode Maintenance
Catalog No. 50792649 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ISAAC Solution is a material specifically formulated for the maintenance of ISAAC reference electrodes, ensuring their optimal performance and longevity.
  • Maintains ISAAC reference electrodes.
  • Supplied as 1 unit per package.
  • Replacement is recommended as required.
  • Contains hazardous material, necessitating special freight handling.
Catalog No. 50-792-649 Supplier Waters Corp Supplier No. 700001949
The ISAAC Solution is a material specifically formulated for the maintenance of ISAAC reference electrodes, ensuring their optimal performance and longevity.
  • Maintains ISAAC reference electrodes.
  • Supplied as 1 unit per package.
  • Replacement is recommended as required.
  • Contains hazardous material, necessitating special freight handling.