Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isatin

Catalog No. 114618100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-461-8100G 100 g
11-461-8500G 500 g
11-461-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-461-8100G Supplier Sigma Aldrich Supplier No. 114618100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isatin

Specifications

CAS 91-56-5
Melting Point 193°C to 195°C (dec.) (lit.)
Molecular Formula C8H5NO2
Linear Formula C8H5NO2
MDL Number MFCD00005718
Quantity 100 g
Synonym 2,3-Diketoindoline; 2,3-Dioxo-2,3-dihydroindole; 2,3-Dioxoindoline; o-Aminobenzoylformic anhydride; Isatic acid lactam; Isatinic acid anhydride; 2,3-Indolinedione; Isatine; NSC 9262
Molecular Weight (g/mol) 147.13
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.