Learn More
TARGETMOL CHEMICALS INC ISATORIBINE 5MG
SDP
Catalog No. 502580826
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Isatoribine(ANA245) free base is a potent TLR7 receptor agonist with anti-hepatitis C virus infection activity for the study of HCV infection. purity: 99%
Catalog No. 50-258-0826 Supplier TARGETMOL CHEMICALS INC Supplier No. T241785MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Isatoribine(ANA245) free base is a potent TLR7 receptor agonist with anti-hepatitis C virus infection activity for the study of HCV infection. purity: 99%