Learn More
Cayman Chemical Isavuconazole 100mg
SDP
Catalog No. 502936876
Encompass_Preferred
A broad-spectrum triazole antifungal agent; inhibits the growth of clinical isolates of A. fumigatus, A. terreus, A. flavus, and A. lentulus (MIC90s = 0.39, 0.39, 2, and 0.25 mg/L, respectively) as well as C. albicans, C. krusei, and C. parapsilosis (MIC50s = 0.03, 0.06, and 0.03 mg/L, respectively); inhibits the growth of C. neoformans and C. gattii clinical isolates (MIC90s = 0.032 and 0.125 mg/L, respectively)
Catalog No. 50-293-6876 Supplier Cayman Chemical Supplier No. 26211100 MG
A broad-spectrum triazole antifungal agent; inhibits the growth of clinical isolates of A. fumigatus, A. terreus, A. flavus, and A. lentulus (MIC90s = 0.39, 0.39, 2, and 0.25 mg/L, respectively) as well as C. albicans, C. krusei, and C. parapsilosis (MIC50s = 0.03, 0.06, and 0.03 mg/L, respectively); inhibits the growth of C. neoformans and C. gattii clinical isolates (MIC90s = 0.032 and 0.125 mg/L, respectively)