Learn More
Medchemexpress LLC Isavuconazole-d4 | 1346598-58-0 | 99.9% | C22H13D4F2N5OS | 1 MG
SDP
Catalog No. 5000355078
Encompass_Preferred
Isavuconazole-d4 (BAL-4815-d4) is a deuterium labeled Isavuconazole (BAL-4815). Isavuconazole is a triazole proagent with antifungal activity against yeasts, molds, and dimorphic fungi. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-55078 Supplier Medchemexpress LLC Supplier No. HY14273S1MG
Isavuconazole-d4 (BAL-4815-d4) is a deuterium labeled Isavuconazole (BAL-4815). Isavuconazole is a triazole proagent with antifungal activity against yeasts, molds, and dimorphic fungi. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.