Learn More
Medchemexpress LLC Isavuconazole-d4 | 1346598-58-0 | 99.9% | C22H13D4F2N5OS | 1 ML
Supplier: Medchemexpress LLC HY14273S10MM/1ML
Isavuconazole-d4 is a deuterium-labeled Isavuconazole, a triazole proagent with antifungal activity against yeasts, molds, and dimorphic fungi. Deuteration, the incorporation of stable heavy isotopes like hydrogen and carbon into drug molecules, is used for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs. The product appears as a solid with a color ranging from light yellow to yellow.
- Can be used as a tracer
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS