Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isavuconazole-d4 | 1346598-58-0 | MFCD32672088 | 99.9% | 441.49 | C22H13D4F2N5OS | 5 MG
SDP

Catalog No. 5000221340
Encompass_Preferred

Isavuconazole-d4 is a deuterium-labeled isotopologue of the antifungal triazole isavuconazole, supplied as a solid tracer/internal standard for analytical and pharmacokinetic applications. It is intended for use in LC-MS/MS assays, quantitative studies, and ADME investigations where an isotopically labeled reference is required.

  • Deuterium-labeled isotopologue suitable as an internal standard.
  • High purity 99.88%.
  • Molecular weight 441.49; formula C22H13D4F2N5OS.
  • Appearance: light yellow to yellow solid.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term).
  • Intended applications: LC-MS/MS, pharmacokinetics, and ADME studies.

Catalog No. 50-002-21340 Supplier Medchemexpress LLC Supplier No. HY14273S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isavuconazole-d4 is a deuterium-labeled isotopologue of the antifungal triazole isavuconazole, supplied as a solid tracer/internal standard for analytical and pharmacokinetic applications. It is intended for use in LC-MS/MS assays, quantitative studies, and ADME investigations where an isotopically labeled reference is required.

  • Deuterium-labeled isotopologue suitable as an internal standard.
  • High purity 99.88%.
  • Molecular weight 441.49; formula C22H13D4F2N5OS.
  • Appearance: light yellow to yellow solid.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term).
  • Intended applications: LC-MS/MS, pharmacokinetics, and ADME studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.