Learn More
Selleck Chemical LLC Isavuconazonium sulfate
SDP
Catalog No. 5000003669
Encompass_Preferred
Isavuconazonium sulfate (BAL8557-002) the prodrug of the active triazole Isavuconazole is an orally active antifungal agent using for invasive aspergillosis and mucormycosis
Catalog No. 50-000-03669 Supplier Selleck Chemical LLC Supplier No. S64585MG
Isavuconazonium sulfate (BAL8557-002) the prodrug of the active triazole Isavuconazole is an orally active antifungal agent using for invasive aspergillosis and mucormycosis